Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAP1A Mouse Monoclonal Antibody

Supplier: Genetex

GTX11264
89357-128EA 448.85 USD InStock
89357-128
Anti-MAP1A Mouse Monoclonal Antibody
Antibodies
Mouse monoclonal [HM-1] to MAP1A

For IHC-Fr: Use at an assay dependent dilution. For WB: Use at a dilution of 1:500. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: MAP1A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty