Anti-Malonaldehyde Deoxyguanosine Adduct Mouse Monoclonal Antibody

Supplier: GeneTex
GTX15163
89360-106EA 560.63 USD
89360-106
Anti-Malonaldehyde Deoxyguanosine Adduct Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Malonaldehyde Deoxyguanosine Adduct
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR