Anti-IgG3 Mouse Monoclonal Antibody [clone: 5G12]

GTX21928
89351-670EA 405.93 USD
89351-670
Anti-IgG3 Mouse Monoclonal Antibody [clone: 5G12]
IgG is a monomeric immunoglobulin, built of two heavy chains gamma and two light chains. Each molecule has two antigen binding sites. This is the most abundant immunoglobulin and is approximately equally distributed in blood and in tissue liquids, constituting 75% of serum immunoglobulins in humans. There are 4 subclasses: IgG1 (66%), IgG2 (23%), IgG3 (7%) and IgG4 (4%).

For ELISA (indirect): Use at a dilution of 1:300,000. Can be used as a coating antibody or as a conjugate. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Secondary
Antigen: IgG3
Clonality: Monoclonal
Clone: 5G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR