Human chorionic gonadotropin (hCG) is a glycoprotein hormone produced by trophoblastic cells of the placenta beginning 10 to 12 days after conception. Maintenance of the fetus in the first trimester of pregnancy requires the production of hCG, which binds to the corpus luteum of the ovary which is stimulated to produce progesterone which in turn maintains the secretory endometrium.
ELISA: Use at an assay dependent dilution. This antibody is suitable for immunochromatography. Can be used as an ELISA pair with GTX21964 and GTX21961. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: CGB
Clonality: Monoclonal
Clone: 28A4
Conjugation: HRP
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human