Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CK14 Mouse Monoclonal Antibody [clone: SPK-14.2]

Supplier: Genetex

GTX72065
89352-946EA 359.26 USD InStock
89352-946
Anti-CK14 Mouse Monoclonal Antibody [clone: SPK-14.2]
Antibodies
Mouse monoclonal [SPK-14.2] to Cytokeratin 14

For IF: Use at a dilution of 1:50-1:100. For WB: Use at a dilution of 1:100-1:200. For FACS: Use at an assay dependent dilution. This antibody is used in research to distinguish stratified epithelial cells from simple epithelial cells and identification of squamous cell carcinomas. The optimal conditions should be determined by the individual laboratory.

Type: Primary
Antigen: CK14
Clonality: Monoclonal
Clone: SPK-14.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty