Anti-CYP1B1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX104424
89350-328EA 557.66 USD
89350-328
Anti-CYP1B1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CYP1B1 (cytochrome P450, family 1, subfamily B, polypeptide 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CYP1B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR