Anti-PPIA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX113520
89357-592EA 557.47 USD
89357-592
Anti-PPIA Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Cyclophilin A (peptidylprolyl isomerase A (cyclophilin A))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PPIA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR