Anti-CNNM3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX115825
89358-432EA 582.76 USD
89358-432
Anti-CNNM3 Rabbit Polyclonal Antibody
Antibodies
Probable metal transporter.

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CNNM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR