Anti-Cyanine Sheep Polyclonal Antibody

Supplier: GeneTex
GTX27627
89362-626EA 659.34 USD
89362-626
Anti-Cyanine Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Cyanine
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR