Anti-CXCR4 Mouse Monoclonal Antibody [clone: 12G5]

GTX72331
89352-994EA 376.85 USD
89352-994
Anti-CXCR4 Mouse Monoclonal Antibody [clone: 12G5]
Mouse Monoclonal to CXCR4 / Fusin / LESTR / HUMSTR

Blocks CD4-dependent infection by certain Ttropic strains of HIV-1. Blocks CD4-independent infection by HIV-2. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CXCR4
Clonality: Monoclonal
Clone: 12G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR