Anti-CXCR1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX100389
89348-454EA 661.1 USD
89348-454
Anti-CXCR1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CXCR1 (chemokine (C-X-C motif) receptor 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CXCR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR