Anti-CXCL13 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX53178
89363-704EA 631.64 USD
89363-704
Anti-CXCL13 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to CXCL13

For WB: Use at a dilution of 1:5,000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CXCL13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR