Anti-CX3CR1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX28020
89362-708EA 632.15 USD
89362-708
Anti-CX3CR1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to CX3CR1

Western blot 1:500 - 50 kD band. Not yet tested in other applications.

Type: Primary
Antigen: CX3CR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR