Anti-CUG-BP Mouse Monoclonal Antibody [clone: 3B1]

Supplier: GeneTex
GTX29549
89362-822EA 644.33 USD
89362-822
Anti-CUG-BP Mouse Monoclonal Antibody [clone: 3B1]
Antibodies
Mouse monoclonal [3B1] to CUG - BP1

For WB: Use at a dilution of 1:500 (ECL). A 50 kDa band can be seen in a Western blot analysis, using HeLa cell lysate. For IF: Use at a dilution of 1:500. Nuclear staining can be seen through immunofluorescence. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CUG-BP
Clonality: Monoclonal
Clone: 3B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR