Anti-CSDA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX77783
89366-302EA 635.46 USD
89366-302
Anti-CSDA Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Antibody to DbpA/CSDA

WB: Suggested dilution at 1 ?g/ml in 5% skim milk / PBS buffer. Optimal dilutions should be determined by end-user.

Type: Primary
Antigen: CSDA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR