Anti-COX1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX113259
89357-410EA 557.66 USD
89357-410
Anti-COX1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to COX1 (prostaglandin-endoperoxide synthase 1 (prostaglandin G/H synthase and cyclooxygenase))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: COX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR