Anti-CTTN Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX11065
89356-414EA 660.25 USD
89356-414
Anti-CTTN Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Cortactin

IF: Use at a dilution of 1/500, this concentration is determined using the mouse fibroblast NIH3T3 cell line. WB: Use at a dilution of 1/1000, this is determined using a rat brain cytosol extract, a whole extract of chicken embryo fibroblasts, or a whole cell extract of the human epidermoid carcinoma A431 cell line. The antibody recognizes human, rat, mouse, and chicken cortactin, (65-75 kDa, 2-3 bands). Staining of cortactin is specifically inhibited with cortactin immunizing peptide. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CTTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR