Anti-COPS4 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX12322
89359-322EA 610.48 USD
89359-322
Anti-COPS4 Rabbit Polyclonal Antibody
Antibodies
CSN4 is a subunit of the JUN activation-domain binding protein 1 (JAB1), containing signalosome complex (or COP9 complex) which has kinase activity and phosphorylates C-JUN. The signalosome complex may play a role in signal transduction.

WB: Use at a dilution of 1/5000 - 1/25000. Predicted molecular weight: 49 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user. Prepare working dilution immediately before use.

Type: Primary
Antigen: COPS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR