Anti-CMKLR1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX71519
89365-276EA 460.78 USD
89365-276
Anti-CMKLR1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal to Human Chemokine - like Receptor 1 ( CMKLR1 )

Immunohistochemistry with formalin fixed paraffin embedded tissues. Not tested for other uses. IHC-P: Use at a dilution of 1/100 - 1/200.

Type: Primary
Antigen: CMKLR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR