Anti-CLMP Goat Polyclonal Antibody

Supplier: GeneTex
GTX47579
89363-630EA 613.32 USD
89363-630
Anti-CLMP Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to CLMP

Preliminary experiments gave an approx 55kDa band in Human Placenta lysates after 1µg/ml antibody staining. Please note that currently we cannot find an explanation in the literature for the band we observe given the calculated size of 41.3kDa according to NP_079045.1. The 55kDa band was successfully blocked by incubation with the immunizing peptide. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates? Have any further splice variants/modified forms been reported?

Type: Primary
Antigen: CLMP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR