Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CLIM1 / PDLIM1 Goat Polyclonal Antibody

Supplier: Genetex

GTX17022
89360-260EA 388.49 USD InStock
89360-260
Anti-CLIM1 / PDLIM1 Goat Polyclonal Antibody
Antibodies
Cytoskeletal protein that may act as an adapter that brings other proteins (like kinases) to the cytoskeleton.

Approx 37kDa band observed in human lung and heart lysates (calculated MW of 36.1kDa according to NP_066272.1). Recommended for use at 0.1-0.3µg/ml.

Type: Primary
Antigen: CLIM1 / PDLIM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty