Anti-CLIC1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX48722
89363-680EA 619.85 USD
89363-680
Anti-CLIC1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to CLIC1

ELISA: 1/312,500. IHC-P: Use at a concentration of 4 - 8 ?g/ml. WB: Use at a concentration of 1.25 ?g/ml. Predicted molecular weight: 27 kDa. Not yet tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CLIC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR