Anti-CLDN7 Rabbit Polyclonal Antibody

GTX30889
89352-746EA 399.78 USD
89352-746
Anti-CLDN7 Rabbit Polyclonal Antibody
Rabbit polyclonal to Claudin 7 (ready-to-use)

Recommended Starting Dilutions:For ICC/IF: Supplied in ready-to-use form. For IHC-P: Supplied in ready-to-use form. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CLDN7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR