Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CLTC Mouse Monoclonal Antibody [clone: 3F133]

Supplier: Genetex

GTX14409
89359-994EA 382.79 USD InStock
89359-994
Anti-CLTC Mouse Monoclonal Antibody [clone: 3F133]
Antibodies
Various cell types express both forms of clathrin LC in different relative levels. LCs are randomly distributed on clathrin triskelion, resulting in four types of clathrin triskelion with different LC composition (3LCa, 2LCa 1LCb, 2LCb-1LCa, 3LCb). In most cells, the LCa form is dominant. In contrast, cells with a regulated secretory pathway contain more LCb than LCa, suggesting that LCb may play a role in the specialized clathrin functions in these cells. Neuronal cells, express splicing variants of both LCa and LCb. A distinguishing feature of the LCb light chain is its ability to be phosphorylated by a casein kinase, at a site which is not present in the LCa sequence.

Recommended Starting Dilutions:ICC: Use at an assay dependent dilution. IF: Use at an assay dependent dilution. IHC-P: Use at a concentration of 1 - 2 μg/ml. IP: Use at an assay dependent dilution. WB: Use at a concentration of 1 - 2 μg/ml. Predicted molecular weight: 28 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CLTC
Clonality: Monoclonal
Clone: 3F133
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty