Anti-KIT Mouse Monoclonal Antibody [clone: 104D2]

Supplier: GeneTex
GTX11289
89357-214EA 492.74 USD
89357-214
Anti-KIT Mouse Monoclonal Antibody [clone: 104D2]
Antibodies
Mouse monoclonal antibody [104D2] to c-Kit

Recommended Starting Dilutions: Flow Cytometry: 1/2 - 1/20. Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: KIT
Clonality: Monoclonal
Clone: 104D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR