Anti-CK1 Mouse Monoclonal Antibody [clone: 3C10F7, 3C10G5, PMC01, 4D12B3,3C10F7C11]

GTX83152
89353-892EA 403.59 USD
89353-892
Anti-CK1 Mouse Monoclonal Antibody [clone: 3C10F7, 3C10G5, PMC01, 4D12B3,3C10F7C11]
Mouse monoclonal antibody [3C10F7, 3C10G5, PMC01, 4D12B3,3C10F7C11] to CK1

Recommended Starting Dilutions:ELISA: 1/10000.WB: 1/500 - 1/2000.IHC: 1/200 - 1/1000.Not yet tested in other applications.Determining optimal working dilutions by titration test.

Type: Primary
Antigen: CK1
Clonality: Monoclonal
Clone: 3C10F7, 3C10G5, PMC01, 4D12B3,3C10F7C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR