Anti-CGA Mouse Monoclonal Antibody [clone: LK2H10]

GTX71948
89352-912EA 341.03 USD
89352-912
Anti-CGA Mouse Monoclonal Antibody [clone: LK2H10]
Mouse Monoclonal to Chromogranin A

Prolonged fixation in buffered formalin can destroy the epitope. The antibody may be used at a dilution of 1:50 to 1:100 with AutoProbe III (GTX28620). Dilute according to the particular application being used. In general, the 0.05M Borate pH 8.0 containing 0.15M Sodium Chloride, 0.05% Sodium Azide, is a good diluent to use with most antibodies. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CGA
Clonality: Monoclonal
Clone: LK2H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR