Anti-CGA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX17064
89351-300EA 688.05 USD
89351-300
Anti-CGA Rabbit Polyclonal Antibody
Antibodies
Chromogranin A (a protein of 439 amino acid which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, phenochromocytomas, paragangliomas, and other neuroendocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.

IHC-P: 1/25 - 1/50. Perform heat mediated antigen retrieval before commencing with IHC staining protocol with 10 mM citrate buffer, pH 6.0. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CGA
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR