Anti-CHRM3 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX71587
89365-292EA 442.73 USD
89365-292
Anti-CHRM3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to mAChR M3

Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CHRM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR