Anti-CHEK1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX100067
89348-188EA 557.66 USD
89348-188
Anti-CHEK1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Chk1 (CHK1 checkpoint homolog (S. pombe))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CHEK1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR