Anti-IgY Goat Polyclonal Antibody

Supplier: GeneTex
GTX77238
89353-568EA 152.47 USD
89353-568
Anti-IgY Goat Polyclonal Antibody
Antibodies
Goats were immunized with highly purified serum IgY in Freunds adjuvant. Serum was collected and the specific antibody isolated by immunoaffinity chromatography and conjugated with fluorescein isothiocyanate (FITC). Goats were immunized with highly purified serum IgY in Freunds adjuvant. Serum was collected and the specific antibody isolated by immunoaffinity chromatography and conjugated with fluorescein isothiocyanate (FITC)

Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR