Goat Anti-Chicken IgM Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Genetex

GTX77008
89353-428EA 154 USD InStock
89353-428
Goat Anti-Chicken IgM Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Alkaline Phosphatase (AP)-conjugated Goat anti-Chicken IgM Polyclonal antibody

Recommended Starting Dilutions: For most applications, use at 1:50 Optimal conjugate working dilutions should be determined for your particular assay conditions.

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR