Anti-IgA Goat Polyclonal Antibody

Supplier: GeneTex

GTX77035
89353-442EA 159.73 USD
89353-442
Anti-IgA Goat Polyclonal Antibody
Antibodies
Goat anti-Chicken IgA Polyclonal antibody

Optimal conjugate working dilutions should be determined for your particular assay conditions.

Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR