Human chorionic gonadotropin (hCG) is a glycoprotein hormone produced by trophoblastic cells of the placenta beginning 10 to 12 days after conception. Maintenance of the fetus in the first trimester of pregnancy requires the production of hCG, which binds to the corpus luteum of the ovary which is stimulated to produce progesterone which in turn maintains the secretory endometrium.
This antibody has been pretitered and quality controlled to work allon formalin-fixed paraffin-embedded as well as acetone fixed cryostat tissue sections. No further titration is required. We suggest an incubation period of 30-60 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation conditions should be determined by the user. Although not required, proteolytic treatment may enhance the immunostaining in overfixed tissues.
Type: Primary
Antigen: CG
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: