Anti-CG Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX29376
89352-610EA 630.94 USD
89352-610
Anti-CG Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to hCG

IHC: This antibody may be diluted to a titer of 1:100-1:500 in an ABC method. Staining Protocol: We suggest an incubation period of 30-60 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation conditions and antibody dilutions should be determined by the user. Although not required, proteolytic treatment may enhance the immunostaining in overfixed tissues.

Type: Primary
Antigen: CG
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR