Anti-CG alpha 6 Mouse Monoclonal Antibody [clone: INN-hCG-72]

Supplier: GeneTex
GTX11387
89357-772EA 581.77 USD
89357-772
Anti-CG alpha 6 Mouse Monoclonal Antibody [clone: INN-hCG-72]
Antibodies
Mouse monoclonal antibody [INN-hCG-72] to CG alpha

Type: Primary
Antigen: CG alpha 6
Clonality: Monoclonal
Clone: INN-hCG-72
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR