Anti-CETP Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX19012
89360-540EA 427.27 USD
89360-540
Anti-CETP Rabbit Polyclonal Antibody
Antibodies
It has been proposed that high density lipoproteins (HDLs) function jointly with lecithin:cholesterol acyltransferase and CETP to facilitate cholesterol transport from tissues to the liver. This mechanism, referred to as reverse cholesterol transport, is physiologically important because it maintains systemic cholesterol levels. CETP is responsible for neutral lipid transfer activity in plasma in numerous species. Since CETP is able to accelerate specifically the exchange of lipid components between pro- and anti-atherogenic lipoprotein fractions, it may be a key determinant of the global atherogenicity of the plasma lipoprotein profile and arises as a possible target in atherosclerosis prevention. CETP has an important role in reverse cholesterol transport and shaping and affecting the composition of plasma lipoproteins. In general elevated levels of CETP have been associated with increased risk of coronary heart disease.

Recommended dilutions: WB: Use at a concentration of 1 - 4 ?g/ml. Predicted molecular weight: 70 kDa. Not yet tested in other applications.Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CETP
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR