Anti-CEA Mouse Monoclonal Antibody [clone: Col-1]

Supplier: GeneTex
GTX17238
89351-356EA 578.85 USD
89351-356
Anti-CEA Mouse Monoclonal Antibody [clone: Col-1]
Antibodies
Carcino Embryonic Antigen (CEA) is synthesised during development in the fetal gut, and is re expressed in increased amounts in intestinal carcinomas and several other tumors. Antibodies to CEA are useful in identifying the origin of various metastatic adenocarcinomas and in distinguishing pulmonary adenocarcinomas (60 to 70% are CEA+) from pleural mesotheliomas (rarely or weakly CEA+).

IHC-P: Use at an assay dependent dilution. IHC-Fr: Use at an assay dependent dilution (acetone fixed sections). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CEA
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR