Anti-CDK9 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX26544
89367-616EA 798.09 USD
89367-616
Anti-CDK9 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CDK9

For ELISA: Use at a dilution of 1:10,000-1:50,000. For IHC-P: Use at a dilution of 1:200-1:1,000. For IP: Use at an assay dependent dilution. For WB: Use at a dilution of 1:500-1:3,000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CDK9
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR