Anti-CDK6 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX77661
89366-228EA 619.97 USD
89366-228
Anti-CDK6 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Antibody to cdk6

WB: Suggested dilution at 1 ?g/ml in 5% skim milk / PBS buffer. Optimal dilutions should be determined by end-user.

Type: Primary
Antigen: CDK6
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR