Anti-CDK5 Mouse Monoclonal Antibody [clone: DC17]

GTX23226
89361-338EA 377.03 USD
89361-338
Anti-CDK5 Mouse Monoclonal Antibody [clone: DC17]
Mouse monoclonal [DC17] to p31 Cdk5

For IF: Use at an assay dependent dilution. For IHC-P: Use at a dilution of 1:200 for 30 min at RT. Staining of formalin fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0, for 10 - 20 min followed by cooling at RT for 20 min. For IP (native and denatured): Use at a dilution of 1:200 of protein lysate (Use protein G). For WB: Use at a dilution of 1:100. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CDK5
Clonality: Monoclonal
Clone: DC17
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR