Anti-CDH13 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX87281
89366-920EA 590.12 USD
89366-920
Anti-CDH13 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CDH13

We recommend the following starting dilutions: WB: 1:500~1:1000 IHC: 1:50~1:100 ELISA: 1:20000Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CDH13
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR