Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDH10 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX87749
89366-946EA 398.53 USD InStock
89366-946
Anti-CDH10 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CDH10

We recommend the following starting dilutions: IF: 1:100~1:500 ELISA: 1:40000Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CDH10
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty