Anti-CDH1 Mouse Monoclonal Antibody [clone: DH01 (DCS-266)]

Supplier: GeneTex
GTX72290
89352-980EA 376.85 USD
89352-980
Anti-CDH1 Mouse Monoclonal Antibody [clone: DH01 (DCS-266)]
Antibodies
Mouse monoclonal [DH01 DCS-266] to Cdh1

For IP (Denatured, Use Protein G): Use at a 1:400 (protein lysate). For WB: Use at a dilution of 1:200. Not tested in other applications. Optimal dilutions should be determined by the researcher.

Type: Primary
Antigen: CDH1
Clonality: Monoclonal
Clone: DH01 (DCS-266)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR