Anti-CDCP1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX82973
89366-760EA 607.87 USD
89366-760
Anti-CDCP1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to CDCP1 (isoform 1: C term)

Approx 90kDa band observed in Human Colon lysates (calculated MW of 92.3kDa according to NP_073753.3). Recommended concentration: 1-3µg/ml. An additional band of unknown identity was also consistently observed at 200kDa. This band was successfully blocked by incubation with the immunising peptide. We would appreciate any feedback from people in the field - have any such results been reported with other antibodies/lysates? Have any further splice variants/modified forms been reported? This antibody is expected to recognize isoform 1 (NP_073753.3) only.

Type: Primary
Antigen: CDCP1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR