Monoclonal Mouse Antibody to CDC47 / MCM7
This antibody may be diluted to a titer of 1:50-1:100 in an ABC method. However, depending on the staining conditions employed, we suggest that final dilution should be determinedby the user. We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation should be determined by the end user. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining. Can also be used for: Immunoprecipitation, Western Blotting
Type: Primary
Antigen: CDC47
Clonality: Monoclonal
Clone: 47DC141
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: