Anti-THY1 Mouse Monoclonal Antibody [clone: F15-42-1]

Supplier: GeneTex
GTX11155
89356-776EA 621.98 USD
89356-776
Anti-THY1 Mouse Monoclonal Antibody [clone: F15-42-1]
Antibodies
Mouse monoclonal antibody [F15-42-1] to CD90

For FACS: Use 10ul of the suggested working dilution to label 106 cells in 100ul. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: THY1
Clonality: Monoclonal
Clone: F15-42-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR