Anti-THY1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX88569
89366-964EA 607.87 USD
89366-964
Anti-THY1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to CD90

Approx 26kDa band observed in Human Brain (Cerebellum) and Human Tonsil lysates (calculated MW of 17.9kDa according to NP_006279.2). The observed molecular weight corresponds to earlier findings with different antibodies from other commercial sources. Recommended concentration: 0.1-0.3µg/ml.

Type: Primary
Antigen: THY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR