Anti-HNRNP-Q Mouse Monoclonal Antibody [clone: I8E4]

GTX10687
89354-812EA 386.22 USD
89354-812
Anti-HNRNP-Q Mouse Monoclonal Antibody [clone: I8E4]
Mouse Monoclonal antibody [I8E4] to hnRNP-Q

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: HNRNP-Q
Clonality: Monoclonal
Clone: I8E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR