Anti-HNRNP-F Mouse Monoclonal Antibody [clone: 3H4]

Supplier: GeneTex

GTX26095
89361-822EA 462.03 USD
89361-822
Anti-HNRNP-F Mouse Monoclonal Antibody [clone: 3H4]
Antibodies
Mouse Monoclonal antibody [3H4] to hnRNP-F

Recommended Starting Dilutions:For WB: Use at 1:500Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: HNRNP-F
Clonality: Monoclonal
Clone: 3H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR